Assay Details
| Gene Symbol: Ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007838.2 | NP_031864.2 | 6 - 7 | 787 | 79 | |
| GenBank mRNA | AK077651.1 | - | 6 - 7 | 649 | 79 | |
| AK081542.1 | - | 6 - 7 | 679 | 79 | ||
| AK084267.1 | - | 6 - 7 | 671 | 79 | ||
| AK148095.1 | - | 6 - 7 | 79 | |||
| AK150683.1 | - | 6 - 7 | 678 | 79 | ||
| BC068132.1 | - | 6 - 7 | 787 | 6808198 | 79 | |
| D89063.1 | - | 6 - 7 | 789 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddost
Genomic Map