Assay Details
| Gene Symbol: Ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007838.2 | NP_031864.2 | 8 - 9 | 1088 | 68 | |
| GenBank mRNA | AK077651.1 | - | 8 - 9 | 950 | 68 | |
| AK081542.1 | - | 8 - 9 | 980 | 68 | ||
| AK084267.1 | - | 8 - 9 | 972 | 68 | ||
| AK148095.1 | - | 8 - 9 | 969 | 68 | ||
| AK150683.1 | - | 8 - 9 | 979 | 68 | ||
| AK177159.1 | - | 1 - 2 | 50 | 68 | ||
| BC068132.1 | - | 8 - 9 | 1088 | 6808198 | 68 | |
| D89063.1 | - | 8 - 9 | 1090 | 68 | ||
More Information
Set Membership: |
|

Ddost
Genomic Map