Assay Details
| Gene Symbol: Dhx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042620.2 | NP_001036085.1 | 5 - 6 | 1273 | 87 | |
| NM_007839.3 | NP_031865.2 | 5 - 6 | 1273 | 87 | ||
| NR_104311.1 | 5 - 6 | 1273 | 87 | |||
| GenBank mRNA | AF017153.1 | - | 5 - 6 | 1121 | 87 | |
| AK145983.1 | - | 5 - 6 | 1223 | 87 | ||
| AK146441.1 | - | 5 - 6 | 1219 | 87 | ||
| AK149869.1 | - | 5 - 6 | 1235 | 87 | ||
| AK167927.1 | - | 5 - 6 | 1146 | 87 | ||
| AK168835.1 | - | 5 - 6 | 1241 | 87 | ||
| AK169174.1 | - | 5 - 6 | 1241 | 87 | ||
| BC003745.1 | - | 5 - 6 | 1215 | 87 | ||
| BC100343.1 | - | 5 - 6 | 1216 | 30935044 | 87 | |
| BC144759.1 | - | 5 - 6 | 1104 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhx15
Genomic Map