Assay Details
| Gene Symbol: Dpagt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007875.2 | NP_031901.2 | 2 - 3 | 478 | 61 | |
| GenBank mRNA | AK164129.1 | - | 2 - 3 | 413 | 61 | |
| BC003943.1 | - | 2 - 3 | 135 | 3487188 | 61 | |
| BC010474.1 | - | 2 - 3 | 478 | 3965178 | 61 | |
| X65603.1 | - | 2 - 3 | 343 | 61 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC003943.1 transcripts.
Set Membership: |
|

Dpagt1
Genomic Map