Assay Details
| Gene Symbol: Dpagt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007875.2 | NP_031901.2 | 8 - 9 | 1359 | 91 | |
| GenBank mRNA | AK008246.1 | - | 7 - 8 | 1211 | 91 | |
| AK008317.1 | - | 7 - 8 | 1212 | 91 | ||
| BC003943.1 | - | 8 - 9 | 1016 | 3487188 | 91 | |
| BC010474.1 | - | 8 - 9 | 1359 | 3965178 | 91 | |
| X65603.1 | - | 8 - 9 | 1224 | 91 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
UDP-N-acetylglucosamine metabolic process
protein N-linked glycosylation
dolichol-linked oligosaccharide biosynthetic process
dolichol metabolic process
dolichol biosynthetic process
UDP-N-acetylglucosamine-dolichyl-phosphate N-acetylglucosaminephosphotransferase activity
UDP-N-acetylglucosamine-lysosomal-enzyme N-acetylglucosaminephosphotransferase activity
phospho-N-acetylmuramoyl-pentapeptide-transferase activity
transferase activity
transferase activity, transferring glycosyl groups
UDP-N-acetylglucosamine metabolic process
protein N-linked glycosylation
dolichol-linked oligosaccharide biosynthetic process
dolichol metabolic process
dolichol biosynthetic process
UDP-N-acetylglucosamine-dolichyl-phosphate N-acetylglucosaminephosphotransferase activity
UDP-N-acetylglucosamine-lysosomal-enzyme N-acetylglucosaminephosphotransferase activity
phospho-N-acetylmuramoyl-pentapeptide-transferase activity
transferase activity
transferase activity, transferring glycosyl groups
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467019 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467018 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467021 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467020 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422208 | X65603.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422208 | AK211438.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422208 | AK212125.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422208 | AK217797.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422208 | BC003943.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422208 | X65603.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422208 | AK211438.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422208 | AK212125.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422208 | AK217797.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422208 | BC003943.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175380 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80040 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239755 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223960 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 133710 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422208 | X65603.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422208 | AK211438.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422208 | AK212125.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422208 | AK217797.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422208 | BC003943.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422208 | X65603.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422208 | AK211438.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422208 | AK212125.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422208 | AK217797.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422208 | BC003943.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422208 | X65603.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422208 | AK211438.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422208 | AK212125.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422208 | AK217797.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422208 | BC003943.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422208 | X65603.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422208 | AK211438.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422208 | AK212125.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422208 | AK217797.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422208 | BC003943.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467028 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01918389 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06769138 | - | - |
| Whole Mouse Genome G4122A | A_51_P422208 | X65603.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422208 | AK211438.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422208 | AK212125.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422208 | AK217797.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422208 | BC003943.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422208 | X65603.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422208 | AK211438.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422208 | AK212125.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422208 | AK217797.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422208 | BC003943.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118411 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06769138 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08132016 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08132025 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467019 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467018 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467020 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26466961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08132029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07520022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07472818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08132036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08167938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08132020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07823326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07520017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08174009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07466779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467023 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10584705 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584709 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584707 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584701 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584702 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584704 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584706 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584708 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448549_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10584700 | - | - |
| Mouse Genome 430 2.0 Array | 1448549_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1448549_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13478_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13478_at | - | - |
| Mouse Expression Set 430 | 1448549_a_at | - | - |
| Mouse Expression Set 430 | 1448549_PM_a_at | - | - |
| Murine Genome U74v2 Set | 103352_PM_at | - | - |
| Murine Genome U74v2 Set | 103352_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448549_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448549_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448549_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448549_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1850324 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001850324 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2641111 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2641111 | - | - |
| mouse-6 v1.1 (Illumina) | scl37040.9_46-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1850324 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37040.9_46-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1850324 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2641111 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2641111 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37040.9_46-S | - | - |

Dpagt1
Genomic Map