Assay Details
| Gene Symbol: Glud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008133.4 | NP_032159.1 | 7 - 8 | 1228 | 118 | |
| GenBank mRNA | AK075971.1 | - | 7 - 8 | 1132 | 118 | |
| AK089152.1 | - | 7 - 8 | 1131 | 118 | ||
| AK150637.1 | - | 7 - 8 | 1228 | 118 | ||
| AK161876.1 | - | 3 - 4 | 362 | 118 | ||
| BC052724.1 | - | 7 - 8 | 1202 | 6812055 | 118 | |
| BC057347.1 | - | 7 - 8 | 1202 | 6812055 | 118 | |
| X57024.1 | - | 7 - 8 | 1133 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glud1
Genomic Map