Assay Details
| Gene Symbol: Gna14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008137.4 | NP_032163.3 | 3 - 4 | 1492 | 63 | |
| GenBank mRNA | AK035320.1 | - | 2 - 3 | 332 | 63 | |
| AK085444.1 | - | 3 - 4 | 1398 | 63 | ||
| AK160808.1 | - | 3 - 4 | 1086 | 63 | ||
| AK168498.1 | - | 3 - 4 | 1148 | 63 | ||
| BC027015.1 | - | 3 - 4 | 960 | 5375817 | 63 | |
| M80631.1 | - | 3 - 4 | 621 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gna14
Genomic Map