Assay Details
| Gene Symbol: Gnat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008141.3 | NP_032167.1 | 5 - 6 | 539 | 61 | |
| GenBank mRNA | AK009388.1 | - | 4 - 5 | 423 | 61 | |
| BC016272.1 | - | 5 - 6 | 537 | 4506728 | 61 | |
| BC085102.1 | - | 3 - 4 | 1286 | 61 | ||
| BC131941.1 | - | 5 - 6 | 534 | 40130218 | 61 | |
| BC131943.1 | - | 5 - 6 | 534 | 40130220 | 61 | |
| L10666.1 | - | 4 - 5 | 461 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnat2
Genomic Map