Assay Details
| Gene Symbol: Gspt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008179.2 | NP_032205.2 | 1 | 491 | 63 | |
| GenBank mRNA | AB003503.1 | - | 1 | 244 | 63 | |
| AK019241.1 | - | 1 | 485 | 63 | ||
| BC117825.1 | - | 1 | 398 | 40092874 | 63 | |
| BC117826.1 | - | 1 | 398 | 40092877 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32539666 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32539663 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32539667 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32539665 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32539664 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32539668 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P647408 | AK050859.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P647408 | AK050859.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301394 | BC117825.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301394 | BC117826.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301394 | AB003503.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301394 | AK019241.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301394 | NM_008179.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301394 | BC117825.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301394 | BC117826.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301394 | AB003503.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301394 | AK019241.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301394 | NM_008179.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494187 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 622542 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132630 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301394 | BC117825.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301394 | BC117826.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301394 | AB003503.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301394 | AK019241.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301394 | NM_008179.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301394 | BC117825.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301394 | BC117826.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301394 | AB003503.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301394 | AK019241.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301394 | NM_008179.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P647408 | AK050859.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P647408 | AK050859.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32539684 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132630 | - | - |
| Whole Mouse Genome G4122A | A_51_P301394 | BC117825.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301394 | BC117826.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301394 | AB003503.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301394 | AK019241.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301394 | NM_008179.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301394 | BC117825.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301394 | BC117826.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301394 | AB003503.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301394 | AK019241.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301394 | NM_008179.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P647408 | AK050859.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P647408 | AK050859.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117419 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132630 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15008697 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15008721 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15008718 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15008684 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32539666 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32539667 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32539664 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32539668 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32539665 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32539663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32539670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15008689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15008729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15008704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14664468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15008710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15008716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15008721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15008722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15081339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539658 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32539653 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10600815 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600818 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600817 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600816 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418109_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455721_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10600814 | - | - |
| Mouse Genome 430 2.0 Array | 1418109_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418109_at | - | - |
| Mouse Genome 430 2.0 Array | 1455721_at | - | - |
| Mouse Genome 430 2.0 Array | 1455721_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14853_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14853_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00005806.mm.1 | NM_008179.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005806.mm.1 | NM_008179.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005806.mm.1 | TC0X00000901.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00005808.mm.1 | NM_008179.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005808.mm.1 | NM_008179.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005808.mm.1 | TC0X00000901.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00005807.mm.1 | NM_008179.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005807.mm.1 | NM_008179.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00005807.mm.1 | TC0X00000901.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418109_at | - | - |
| Mouse Expression Set 430 | 1418109_PM_at | - | - |
| Mouse Expression Set 430 | 1455721_PM_at | - | - |
| Mouse Expression Set 430 | 1455721_at | - | - |
| Murine Genome U74v2 Set | 103261_PM_at | - | - |
| Murine Genome U74v2 Set | 103261_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418109_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418109_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418109_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418109_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455721_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455721_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 625624 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2100010 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2100010 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100540026 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002100010 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2671253 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2671253 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248661 | - | - |
| mouse-6 v1.1 (Illumina) | scl54777.3.514_32-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0014853.2_151-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100540026 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2100010 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0014853.2_151-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100540026 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100010 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2671253 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2671253 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0014853.2_151-S | - | - |

Gspt2
Genomic Map