Assay Details
| Gene Symbol: Icam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008319.2 | NP_032345.2 | 7 - 8 | 1801 | 70 | |
| GenBank mRNA | AK135040.1 | - | 7 - 8 | 1714 | 70 | |
| BC113140.1 | - | 7 - 8 | 1749 | 40048010 | 70 | |
| U06483.1 | - | 7 - 8 | 1749 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Icam5
Genomic Map