Assay Details
| Gene Symbol: Junb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044253.1 | - | 823 | 140 | ||
| AK088643.1 | - | 1 | 823 | 140 | ||
| AK159083.1 | - | 1 | 822 | 140 | ||
| AK159520.1 | - | 1 | 822 | 140 | ||
| AK159661.1 | - | 1 | 824 | 140 | ||
| BC003790.1 | - | 1 | 801 | 3592704 | 140 | |
| BC092302.1 | - | 1 | 799 | 6399302 | 140 | |
| J03236.1 | - | 1 | 852 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Citations (1)
GATA-3 directly remodels the IL-10 locus independently of IL-4 in CD4+ T cells.
- Authors
- Shoemaker J, Saraiva M, O'Garra A
- Journal
- J Immunol - Page No 3470 (Wed Mar 15 00:00:00 EST 2006)
- Volume (Year)
- 176 (2006)
- Product Usage
- -

Junb
Genomic Map