Assay Details
Gene Symbol: Klf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008453.5 | NP_032479.1 | 3 - 4 | 851 | 110 | |
GenBank mRNA | AK007959.1 | - | 3 - 4 | 850 | 110 | |
AK010713.1 | - | 3 - 4 | 796 | 110 | ||
AK046659.1 | - | 2 - 3 | 1467 | 110 | ||
AK081467.1 | - | 1 - 2 | 335 | 110 | ||
AK143838.1 | - | 3 - 4 | 854 | 110 | ||
AK157576.1 | - | 3 - 4 | 986 | 109 | ||
BC116938.1 | - | 3 - 4 | 818 | 40126257 | 110 | |
BC119214.1 | - | 3 - 4 | 818 | 8733963 | 110 | |
DQ981866.1 | - | 3 - 4 | 746 | 110 | ||
U36340.1 | - | 2 - 3 | 587 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
