Assay Details
| Gene Symbol: Gzmm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB015728.1 | - | 1 - 2 | 451 | 111 | |
| BC139217.1 | - | 1 - 2 | 320 | 8862239 | 111 | |
| BC139218.1 | - | 1 - 2 | 320 | 8862240 | 111 | |
| BG082217.2 | - | 1 - 2 | 83 | 111 | ||
| L76741.1 | - | 1 - 2 | 60 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gzmm
Genomic Map