Assay Details
| Gene Symbol: St3gal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161774.2 | NP_001155246.1 | 7 - 8 | 698 | 94 | |
| NM_001285520.1 | NP_001272449.1 | 7 - 8 | 1028 | 94 | ||
| NM_001285521.1 | NP_001272450.1 | 6 - 7 | 650 | 94 | ||
| NM_009176.5 | NP_033202.3 | 7 - 8 | 1042 | 94 | ||
| GenBank mRNA | AK005053.1 | - | 7 - 8 | 987 | 94 | |
| AK013016.1 | - | 6 - 7 | 589 | 94 | ||
| AK136442.1 | - | 7 - 8 | 979 | 94 | ||
| BC006710.1 | - | 7 - 8 | 666 | 3584323 | 94 | |
| X84234.1 | - | 7 - 8 | 521 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

St3gal3
Genomic Map