Assay Details
Gene Symbol: Taf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009315.3 | NP_033341.1 | 11 - 12 | 1233 | 65 | |
GenBank mRNA | AK047944.1 | - | 1 - 2 | 349 | 65 | |
AK153731.1 | - | 1281 | 65 | |||
AK154867.1 | - | 11 - 12 | 1267 | 65 | ||
BC058583.1 | - | 11 - 12 | 1233 | 6514960 | 65 | |
D49439.1 | - | 10 - 11 | 1166 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
