Assay Details
Gene Symbol: Taf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009315.3 | NP_033341.1 | 13 - 14 | 1541 | 55 | |
GenBank mRNA | AK154867.1 | - | 13 - 14 | 1575 | 55 | |
AK204792.1 | - | 1 - 2 | 120 | 55 | ||
BC058583.1 | - | 13 - 14 | 1541 | 6514960 | 55 | |
D49439.1 | - | 12 - 13 | 1474 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
negative regulation of cell proliferation
negative regulation of cell cycle
regulation of sequence-specific DNA binding transcription factor activity
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein heterodimerization activity
transcription, DNA-templated
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
negative regulation of cell proliferation
negative regulation of cell cycle
regulation of sequence-specific DNA binding transcription factor activity
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993109 | BC058583.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993109 | D49439.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993109 | AK154867.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993109 | NM_009315.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 592770 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 746598 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801895 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704405 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 911136 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107386 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993109 | BC058583.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993109 | D49439.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993109 | AK154867.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993109 | NM_009315.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443053 | BC058583.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443053 | D49439.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443053 | AK153731.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443053 | AK154867.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443053 | NM_009315.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129241 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129235 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129236 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129244 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129265 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129254 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129259 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129243 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129262 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129252 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129263 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129249 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129255 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129237 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129239 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24129242 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01232708 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01232716 | - | - |
Whole Mouse Genome G4122A | A_51_P443053 | BC058583.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P443053 | D49439.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P443053 | AK153731.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P443053 | AK154867.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P443053 | NM_009315.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124575 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04187701 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04187696 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04187700 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04187697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129243 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129225 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04698078 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04187701 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04698080 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129265 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04829777 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04346511 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129254 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129215 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04477890 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04829775 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129263 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129244 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04346508 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129229 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129262 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129217 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04300621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129239 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129211 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129228 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129250 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129214 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129259 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129248 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129220 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04829772 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129257 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129255 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04829776 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129241 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24129252 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10535000 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534998 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10535005 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534995 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10535004 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534994 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534991 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534997 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534993 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534996 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534992 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10535001 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10535003 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10534999 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10535002 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418593_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10534990 | - | - |
Mouse Genome 430 2.0 Array | 1418593_at | - | - |
Mouse Genome 430 2.0 Array | 1418593_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21343_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21343_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500031683.mm.1 | TC0500003474.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500031683.mm.1 | NM_009315.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500031683.mm.1 | NM_009315.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500031685.mm.1 | TC0500003474.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500031685.mm.1 | NM_009315.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500031685.mm.1 | NM_009315.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500031684.mm.1 | TC0500003474.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500031684.mm.1 | NM_009315.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500031684.mm.1 | NM_009315.3 | Refseq |
Mouse Expression Set 430 | 1418593_at | - | - |
Mouse Expression Set 430 | 1418593_PM_at | - | - |
Murine Genome U74v2 Set | 103591_PM_at | - | - |
Murine Genome U74v2 Set | 103591_at | - | - |
Mouse Genome 430A 2.0 Array | 1418593_at | - | - |
Mouse Genome 430A 2.0 Array | 1418593_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418593_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418593_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4280497 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2470541 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4280497 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002470541 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004280497 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1241137 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252302 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1241137 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2475184 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1252302 | - | - |
mouse-6 v1.1 (Illumina) | scl0000117.1_15-S | - | - |
mouse-6 v1.1 (Illumina) | scl25859.15.1_31-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4280497 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2470541 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl25859.15.1_31-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0000117.1_15-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280497 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2470541 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1252302 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1241137 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1241137 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1252302 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0000117.1_15-S | - | - |
