Assay Details
| Gene Symbol: Tcf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009331.3 | NP_033357.1 | 3 - 4 | 257 | 88 | |
| GenBank mRNA | AK037616.1 | - | 3 - 4 | 258 | 88 | |
| BC056630.1 | - | 2 - 3 | 499 | 88 | ||
| BC138596.1 | - | 2 - 3 | 237 | 8861616 | 88 | |
| BC145343.1 | - | 2 - 3 | 237 | 9053899 | 88 | |
| X61385.1 | - | 4 - 5 | 399 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tcf7
Genomic Map