Assay Details
| Gene Symbol: Tcf7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079822.2 | NP_001073290.1 | 9 - 10 | 1114 | 105 | |
| NM_009332.3 | NP_033358.2 | 8 - 9 | 1072 | 105 | ||
| GenBank mRNA | AF057691.1 | - | 4 - 5 | 460 | 105 | |
| AJ223069.1 | - | 8 - 9 | 980 | 105 | ||
| AK052721.1 | - | 8 - 9 | 1282 | 105 | ||
| BC128305.1 | - | 9 - 10 | 1021 | 40109611 | 105 | |
| BC128306.1 | - | 8 - 9 | 979 | 40109613 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tcf7l1
Genomic Map