Assay Details
| Gene Symbol: Tph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136084.2 | NP_001129556.1 | 4 - 5 | 535 | 90 | |
| NM_009414.3 | NP_033440.1 | 4 - 5 | 846 | 90 | ||
| GenBank mRNA | AF273257.1 | - | 4 - 5 | 536 | 90 | |
| AK146134.1 | - | 4 - 5 | 535 | 90 | ||
| AK146171.1 | - | 4 - 5 | 535 | 90 | ||
| BC072582.1 | - | 4 - 5 | 846 | 5718460 | 90 | |
| J04758.1 | - | 4 - 5 | 506 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Citations (1)
Homeodomain transcription factor NKX2.2 functions in immature cells to control enteroendocrine differentiation and is expressed in gastrointestinal neuroendocrine tumors.
- Authors
- Wang YC, Gallego-Arteche E, Iezza G, Yuan X, Matli MR, Choo SP, Zuraek MB, Gogia R, Lynn FC, German MS, Bergsland EK, Donner DB, Warren RS, Nakakura EK
- Journal
- Endocr Relat Cancer - Page No 267 (Sun Mar 01 00:00:00 EST 2009)
- Volume (Year)
- 16 (2009)
- Product Usage
- -

Tph1
Genomic Map