Assay Details
Gene Symbol: Crisp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204071.1 | NP_001191000.1 | 6 - 7 | 508 | 112 | |
NM_009420.2 | NP_033446.1 | 6 - 7 | 526 | 112 | ||
GenBank mRNA | BC049615.1 | - | 6 - 7 | 478 | 6742033 | 112 |
M25533.1 | - | 6 - 7 | 478 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
