Assay Details
| Gene Symbol: Traf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009423.4 | NP_033449.2 | 5 - 6 | 731 | 64 | |
| GenBank mRNA | AF233449.1 | - | 5 - 6 | 630 | 64 | |
| AK035056.1 | - | 5 - 6 | 709 | 64 | ||
| AK040877.1 | - | 709 | 64 | |||
| AK046015.1 | - | 5 - 6 | 771 | 64 | ||
| AK053900.1 | - | 5 - 6 | 728 | 64 | ||
| BC020455.1 | - | 5 - 6 | 716 | 64 | ||
| X92346.1 | - | 5 - 6 | 751 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Traf4
Genomic Map