Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 10 - 11 | 976 | 73 | |
| GenBank mRNA | AB008516.1 | - | 9 - 10 | 853 | 73 | |
| AK053765.1 | - | 10 - 11 | 1080 | 73 | ||
| AK088273.1 | - | 9 - 10 | 923 | 73 | ||
| AK220495.1 | - | 10 - 11 | 1018 | 73 | ||
| BC019173.1 | - | 10 - 11 | 945 | 73 | ||
| BC037654.1 | - | 9 - 10 | 878 | 73 | ||
| BC049277.1 | - | 9 - 10 | 915 | 73 | ||
| BC057207.1 | - | 9 - 10 | 880 | 73 | ||
| BC145489.1 | - | 9 - 10 | 893 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc3
Genomic Map