Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 19 - 20 | 1789 | 66 | |
| GenBank mRNA | AB008516.1 | - | 18 - 19 | 1666 | 66 | |
| AK053765.1 | - | 19 - 20 | 1893 | 66 | ||
| AK084201.1 | - | 9 - 10 | 824 | 66 | ||
| AK088273.1 | - | 18 - 19 | 1736 | 66 | ||
| AK165850.1 | - | 8 - 9 | 697 | 66 | ||
| AK166120.1 | - | 9 - 10 | 729 | 66 | ||
| AK166164.1 | - | 10 - 11 | 878 | 66 | ||
| AK220495.1 | - | 19 - 20 | 1831 | 66 | ||
| BC019173.1 | - | 19 - 20 | 1758 | 66 | ||
| BC037654.1 | - | 18 - 19 | 1691 | 66 | ||
| BC057207.1 | - | 18 - 19 | 1693 | 66 | ||
| BC145489.1 | - | 14 - 15 | 1309 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc3
Genomic Map