Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 35 - 36 | 4695 | 71 | |
| GenBank mRNA | AB008516.1 | - | 34 - 35 | 4572 | 71 | |
| AK148494.1 | - | 8 - 9 | 1719 | 71 | ||
| AK164106.1 | - | 3 - 4 | 296 | 71 | ||
| AK164492.1 | - | 3 - 4 | 1217 | 71 | ||
| BC145489.1 | - | 30 - 31 | 4215 | 71 | ||
More Information
Set Membership: |
|

Ttc3
Genomic Map