Assay Details
| Gene Symbol: Ucp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009464.3 | NP_033490.1 | 2 - 3 | 322 | 75 | |
| GenBank mRNA | AB008216.1 | - | 1 - 2 | 125 | 75 | |
| AB010742.1 | - | 2 - 3 | 322 | 75 | ||
| AF030164.1 | - | 1 - 2 | 163 | 75 | ||
| AF032902.1 | - | 2 - 3 | 289 | 75 | ||
| AF053352.1 | - | 2 - 3 | 294 | 75 | ||
| BC055901.1 | - | 2 - 3 | 319 | 75 | ||
| BC139430.1 | - | 1 - 2 | 190 | 8862452 | 75 | |
| BC139431.1 | - | 1 - 2 | 190 | 8862453 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Fibroblast growth factor 21 corrects obesity in mice.
- Authors
- Coskun T, Bina HA, Schneider MA, Dunbar JD, Hu CC, Chen Y, Moller DE, Kharitonenkov A
- Journal
- Endocrinology - Page No 6018 (Mon Dec 01 00:00:00 EST 2008)
- Volume (Year)
- 149 (2008)
- Product Usage
- -

Ucp3
Genomic Map