Assay Details
| Gene Symbol: Vhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009507.3 | NP_033533.1 | 1 - 2 | 358 | 83 | |
| GenBank mRNA | AK159238.1 | - | 1 - 2 | 379 | 83 | |
| AK161408.1 | - | 1 - 2 | 355 | 83 | ||
| AK171264.1 | - | 1 - 2 | 360 | 83 | ||
| AK171604.1 | - | 1 - 2 | 368 | 83 | ||
| BC052417.1 | - | 1 - 2 | 357 | 6402536 | 83 | |
| S76748.1 | - | 1 - 2 | 358 | 83 | ||
| U12570.1 | - | 1 - 2 | 374 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vhl
Genomic Map