Assay Details
| Gene Symbol: Dnajc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009584.4 | NP_033610.1 | 11 - 12 | 1290 | 70 | |
| GenBank mRNA | AK131964.1 | - | 11 - 12 | 1311 | 70 | |
| AK167991.1 | - | 11 - 12 | 1311 | 70 | ||
| BC021755.1 | - | 11 - 12 | 1295 | 70 | ||
| BC052027.1 | - | 11 - 12 | 1317 | 5718637 | 70 | |
| D63784.1 | - | 11 - 12 | 1289 | 70 | ||
| U53208.1 | - | 11 - 12 | 1245 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc2
Genomic Map