Assay Details
| Gene Symbol: Dpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159543.1 | NP_001153015.1 | 23 - 24 | 2471 | 92 | |
| NM_010074.3 | NP_034204.1 | 23 - 24 | 2471 | 92 | ||
| GenBank mRNA | AK085370.1 | - | 23 - 24 | 2054 | 92 | |
| AK153939.1 | - | 23 - 24 | 2085 | 92 | ||
| AK163085.1 | - | 23 - 24 | 2476 | 92 | ||
| AK164775.1 | - | 23 - 24 | 2054 | 92 | ||
| BC022183.1 | - | 24 - 25 | 2333 | 3982742 | 92 | |
| X58384.1 | - | 23 - 24 | 2148 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpp4
Genomic Map