Assay Details
Gene Symbol: Prl8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289919.1 | NP_001276848.1 | 4 - 5 | 410 | 86 | |
NM_010088.2 | NP_034218.1 | 4 - 5 | 413 | 86 | ||
GenBank mRNA | AF011385.1 | - | 4 - 5 | 397 | 86 | |
AF015729.1 | - | 4 - 5 | 407 | 86 | ||
AK131668.1 | - | 4 - 5 | 411 | 86 | ||
AK146049.1 | - | 4 - 5 | 414 | 86 | ||
BC061150.1 | - | 4 - 5 | 410 | 30317747 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
