Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800008339.mm.1 |
NM_001113384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008339.mm.1 |
NM_001113384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008339.mm.1 |
NM_010308.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008339.mm.1 |
NM_010308.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008339.mm.1 |
TC0800001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800008340.mm.1 |
NM_001113384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008340.mm.1 |
NM_001113384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008340.mm.1 |
NM_010308.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008340.mm.1 |
NM_010308.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008340.mm.1 |
TC0800001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800008341.mm.1 |
NM_001113384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008341.mm.1 |
NM_001113384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008341.mm.1 |
NM_010308.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008341.mm.1 |
NM_010308.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008341.mm.1 |
TC0800001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800008342.mm.1 |
NM_001113384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008342.mm.1 |
NM_001113384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008342.mm.1 |
NM_010308.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008342.mm.1 |
NM_010308.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008342.mm.1 |
TC0800001085.mm.1 |
MTA TC |