Assay Details
| Gene Symbol: Gnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010321.1 | NP_034451.1 | 1 - 2 | 222 | 63 | |
| GenBank mRNA | BC014283.1 | - | 1 - 2 | 221 | 4210236 | 63 |
| BC087960.1 | - | 1 - 2 | 222 | 6766468 | 63 | |
| D89664.1 | - | 1 - 2 | 222 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnmt
Genomic Map