Assay Details
| Gene Symbol: Gpx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010343.2 | NP_034473.2 | 1 - 2 | 115 | 64 | |
| GenBank mRNA | AK029731.1 | - | 1 - 2 | 116 | 64 | |
| BC100749.1 | - | 1 - 2 | 115 | 40046457 | 64 | |
| BC103588.1 | - | 1 - 2 | 115 | 40045838 | 64 | |
| BC103589.1 | - | 1 - 2 | 115 | 40045837 | 64 | |
| BC105648.1 | - | 1 - 2 | 115 | 40046458 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpx5
Genomic Map