Assay Details
| Gene Symbol: Grb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010346.2 | NP_034476.1 | 9 - 10 | 1438 | 97 | |
| GenBank mRNA | AK057640.1 | - | 9 - 10 | 1431 | 97 | |
| AK145996.1 | - | 9 - 10 | 1254 | 97 | ||
| AK168438.1 | - | 9 - 10 | 1222 | 97 | ||
| AK220277.1 | - | 8 - 9 | 1329 | 97 | ||
| BC003295.1 | - | 9 - 10 | 1181 | 3583896 | 97 | |
| BC081547.1 | - | 8 - 9 | 1427 | 97 | ||
| M94450.1 | - | 9 - 10 | 1460 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grb7
Genomic Map