Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500030305.mm.1 |
TC0500003369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030305.mm.1 |
NM_001080746.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030305.mm.1 |
NM_001080747.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030305.mm.1 |
NM_001080748.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030305.mm.1 |
NM_001080749.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030305.mm.1 |
NM_010365.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030306.mm.1 |
TC0500003369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030306.mm.1 |
NM_001080746.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030306.mm.1 |
NM_001080747.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030306.mm.1 |
NM_001080748.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030306.mm.1 |
NM_001080749.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030306.mm.1 |
NM_010365.4 |
Refseq |