Assay Details
| Gene Symbol: Jund | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286944.1 | NP_001273873.1 | 1 | 1535 | 106 | |
| NM_010592.5 | NP_034722.1 | 1 | 1535 | 106 | ||
| GenBank mRNA | BC010572.1 | - | 1 | 332 | 3583807 | 106 |
| BC023211.1 | - | 1 | 326 | 106 | ||
| BC094068.1 | - | 1 | 316 | 4456297 | 106 | |
| BE648283.1 | - | 1 | 106 | |||
| CK621709.1 | - | 1 | 323 | 106 | ||
| J05205.1 | - | 1 | 259 | 106 | ||
| X15358.1 | - | 1 | 248 | 106 | ||
| Gene Symbol: Gm11175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK135112.1 | - | 1 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Jund
Genomic Map