Assay Details
| Gene Symbol: Kcnh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038607.2 | NP_001033696.1 | 3 - 4 | 540 | 118 | |
| NM_010600.3 | NP_034730.1 | 3 - 4 | 540 | 118 | ||
| GenBank mRNA | AK140193.1 | - | 3 - 4 | 446 | 118 | |
| AK147461.1 | - | 3 - 4 | 534 | 118 | ||
| BC109012.1 | - | 3 - 4 | 399 | 40048708 | 118 | |
| BC109013.1 | - | 3 - 4 | 399 | 40048709 | 118 | |
| U04294.1 | - | 3 - 4 | 422 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnh1
Genomic Map