Assay Details
| Gene Symbol: Lbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010692.3 | NP_034822.1 | 1 - 2 | 262 | 105 | |
| GenBank mRNA | AF146150.1 | - | 1 - 2 | 203 | 105 | |
| AK002897.1 | - | 1 - 2 | 262 | 105 | ||
| BC109347.2 | - | 1 - 2 | 257 | 40055655 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591363 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591351 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591362 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591360 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591352 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591361 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591350 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165602 | NM_010692.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165602 | AF146150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165602 | AK002897.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165602 | NM_010692.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165602 | AF146150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165602 | AK002897.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 950483 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165602 | NM_010692.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165602 | AF146150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165602 | AK002897.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165602 | NM_010692.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165602 | AF146150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165602 | AK002897.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165602 | NM_010692.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165602 | AF146150.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165602 | AK002897.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165602 | NM_010692.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165602 | AF146150.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165602 | AK002897.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591322 | - | - |
| Whole Mouse Genome G4122A | A_51_P165602 | NM_010692.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165602 | AF146150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165602 | AK002897.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165602 | NM_010692.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165602 | AF146150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165602 | AK002897.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101993 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591351 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591350 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591363 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591360 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591361 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591352 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591341 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10539317 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539318 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539319 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420714_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539316 | - | - |
| Mouse Genome 430 2.0 Array | 1420714_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420714_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16815_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16815_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600006917.mm.1 | TC0600000920.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600006917.mm.1 | NM_010692.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006917.mm.1 | NM_010692.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006919.mm.1 | TC0600000920.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600006919.mm.1 | NM_010692.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006919.mm.1 | NM_010692.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006920.mm.1 | TC0600000920.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600006920.mm.1 | NM_010692.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006920.mm.1 | NM_010692.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006921.mm.1 | TC0600000920.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600006921.mm.1 | NM_010692.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006921.mm.1 | NM_010692.3 | Refseq |
| Mouse Expression Set 430 | 1420714_PM_at | - | - |
| Mouse Expression Set 430 | 1420714_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420714_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420714_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420714_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420714_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 829373 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1450280 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1450280 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001450280 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1258787 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1258787 | - | - |
| mouse-6 v1.1 (Illumina) | scl29848.3.1_62-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1450280 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29848.3.1_62-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1450280 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1258787 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1258787 | - | - |

Lbx2
Genomic Map