Assay Details
| Gene Symbol: Lgals9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159301.1 | NP_001152773.1 | 3 - 4 | 437 | 86 | |
| NM_010708.2 | NP_034838.2 | 3 - 4 | 437 | 86 | ||
| GenBank mRNA | AK146044.1 | - | 3 - 4 | 393 | 86 | |
| BC003754.1 | - | 3 - 4 | 364 | 3601419 | 86 | |
| CA458877.1 | - | 3 - 4 | 427 | 86 | ||
| U55060.1 | - | 3 - 4 | 363 | 86 | ||
| U55061.1 | - | 3 - 4 | 344 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lgals9
Genomic Map