Assay Details
Gene Symbol: Osmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011019.3 | NP_035149.2 | 5 - 6 | 818 | 99 | |
GenBank mRNA | AB015978.1 | - | 5 - 6 | 1472 | 99 | |
AF058805.1 | - | 5 - 6 | 788 | 99 | ||
AK087179.1 | - | 5 - 6 | 776 | 99 | ||
BC137703.1 | - | 4 - 5 | 713 | 8860722 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
