Assay Details
| Gene Symbol: Tbxas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011539.3 | NP_035669.3 | 6 - 7 | 709 | 66 | |
| GenBank mRNA | AK048544.1 | - | 6 - 7 | 691 | 66 | |
| AK085414.1 | - | 3 - 4 | 460 | 66 | ||
| AK139479.1 | - | 6 - 7 | 672 | 66 | ||
| BC029014.1 | - | 6 - 7 | 711 | 5007629 | 66 | |
| L18868.1 | - | 6 - 7 | 580 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbxas1
Genomic Map