Assay Details
Gene Symbol: Trim28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011588.3 | NP_035718.2 | 9 - 10 | 1893 | 66 | |
GenBank mRNA | AF230391.1 | - | 9 - 10 | 1494 | 66 | |
AF230392.1 | - | 9 - 10 | 1494 | 66 | ||
AK141357.1 | - | 9 - 10 | 1637 | 66 | ||
AK149995.1 | - | 6 - 7 | 718 | 66 | ||
BC020452.1 | - | 9 - 10 | 1324 | 66 | ||
BC023335.1 | - | 9 - 10 | 1602 | 66 | ||
BC058391.1 | - | 9 - 10 | 1339 | 6817165 | 66 | |
BC089337.1 | - | 9 - 10 | 1893 | 6811294 | 66 | |
U67303.1 | - | 9 - 10 | 1884 | 66 | ||
X99644.1 | - | 9 - 10 | 1494 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
