Assay Details
Gene Symbol: Zrsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011663.3 | NP_035793.1 | 1 | 1689 | 62 | |
GenBank mRNA | AK030552.1 | - | 1 | 479 | 62 | |
BC132555.1 | - | 2 | 747 | 40130832 | 62 | |
BC138594.1 | - | 1 | 757 | 8861614 | 62 | |
D17407.1 | - | 2 | 1634 | 62 | ||
S69507.1 | - | 1 | 1623 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643663 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643657 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643661 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643662 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643659 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643655 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643658 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643656 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643660 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407675 | D17407.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407675 | S69507.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407675 | NM_011663.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407675 | AK030552.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407675 | D17407.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407675 | S69507.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407675 | NM_011663.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407675 | AK030552.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102717 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407675 | D17407.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407675 | S69507.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407675 | NM_011663.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407675 | AK030552.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407675 | D17407.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407675 | S69507.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407675 | NM_011663.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407675 | AK030552.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P407675 | D17407.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P407675 | S69507.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P407675 | NM_011663.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P407675 | AK030552.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P407675 | D17407.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P407675 | S69507.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P407675 | NM_011663.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P407675 | AK030552.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139874 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09127885 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09127884 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09127883 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643658 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643657 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643663 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643660 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643655 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643659 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643656 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643662 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27643661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27643645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643639 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643637 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643664 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09362726 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09620743 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643672 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643641 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643666 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643665 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09670934 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643649 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643676 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09620738 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643657 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09362727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09127885 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643661 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09362730 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643633 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643631 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643635 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643653 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09362731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643628 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643640 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643655 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09127894 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27643652 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09366035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08989696 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10374561 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10374562 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384666 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449354_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10374560 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10384662 | - | - |
Mouse Genome 430 2.0 Array | 1449354_at | - | - |
Mouse Genome 430 2.0 Array | 1449354_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22183_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22183_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100001857.mm.1 | NM_011663.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100001857.mm.1 | NM_011663.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100001857.mm.1 | TC1100000231.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100001858.mm.1 | NM_011663.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100001858.mm.1 | NM_011663.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100001858.mm.1 | TC1100000231.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100001861.mm.1 | NM_011663.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100001861.mm.1 | NM_011663.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100001861.mm.1 | TC1100000231.mm.1 | MTA TC |
Mouse Expression Set 430 | 1449354_PM_at | - | - |
Mouse Expression Set 430 | 1449354_at | - | - |
Murine Genome U74v2 Set | 102817_at | - | - |
Murine Genome U74v2 Set | 102817_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449354_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449354_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449354_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449354_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 730858 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4010463 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004010463 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2448615 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2448615 | - | - |
mouse-6 v1.1 (Illumina) | scl022183.2_94-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4010463 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl022183.2_94-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4010463 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2448615 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2448615 | - | - |
