Assay Details
| Gene Symbol: Uchl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011670.2 | NP_035800.2 | 5 - 6 | 574 | 68 | |
| GenBank mRNA | AB025313.1 | - | 5 - 6 | 425 | 68 | |
| AF172334.1 | - | 5 - 6 | 413 | 68 | ||
| AK004066.1 | - | 5 - 6 | 451 | 68 | ||
| AK013729.1 | - | 5 - 6 | 454 | 68 | ||
| AK170728.1 | - | 3 - 4 | 412 | 68 | ||
| BC039177.1 | - | 5 - 6 | 440 | 4506229 | 68 | |
| JX050004.1 | - | 3 - 4 | 378 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uchl1
Genomic Map