Assay Details
| Gene Symbol: Sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110791.1 | NP_001104261.1 | 9 - 10 | 1463 | 79 | |
| NM_011750.2 | NP_035880.2 | 9 - 10 | 1463 | 79 | ||
| NR_122118.1 | 8 - 9 | 1387 | 79 | |||
| GenBank mRNA | AK133895.1 | - | 9 - 10 | 1252 | 79 | |
| AK146151.1 | - | 9 - 10 | 1224 | 79 | ||
| AK147081.1 | - | 9 - 10 | 1232 | 79 | ||
| AK154705.1 | - | 9 - 10 | 1231 | 79 | ||
| AK157845.1 | - | 9 - 10 | 1232 | 79 | ||
| BC009091.1 | - | 9 - 10 | 1208 | 3157495 | 79 | |
| BC055370.1 | - | 9 - 10 | 1222 | 30055109 | 79 | |
| CD563870.1 | - | 3 - 4 | 324 | 79 | ||
| X80159.1 | - | 1 - 2 | 180 | 79 | ||
| X85802.1 | - | 9 - 10 | 1220 | 79 | ||
More Information
Set Membership: |
|

Sf1
Genomic Map