Assay Details
| Gene Symbol: Impdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011830.3 | NP_035960.2 | 2 - 3 | 180 | 129 | |
| GenBank mRNA | AK002676.1 | - | 2 - 3 | 212 | 129 | |
| AK011777.1 | - | 2 - 3 | 213 | 129 | ||
| AK143505.1 | - | 2 - 3 | 221 | 129 | ||
| AK146362.1 | - | 2 - 3 | 246 | 129 | ||
| AK150713.1 | - | 2 - 3 | 216 | 129 | ||
| AK151233.1 | - | 2 - 3 | 216 | 129 | ||
| AK151706.1 | - | 2 - 3 | 239 | 129 | ||
| AK152691.1 | - | 2 - 3 | 213 | 129 | ||
| AK160569.1 | - | 2 - 3 | 213 | 129 | ||
| AK166667.1 | - | 2 - 3 | 215 | 129 | ||
| AK169129.1 | - | 2 - 3 | 246 | 129 | ||
| AK169788.1 | - | 2 - 3 | 246 | 129 | ||
| BC010314.1 | - | 2 - 3 | 180 | 2646679 | 129 | |
| BC052671.1 | - | 2 - 3 | 201 | 30058399 | 129 | |
| M33934.1 | - | 2 - 3 | 213 | 129 | ||
| M98333.1 | - | 2 - 3 | 147 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Impdh2
Genomic Map