Assay Details
| Gene Symbol: Impdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011830.3 | NP_035960.2 | 8 - 9 | 946 | 83 | |
| GenBank mRNA | AK143505.1 | - | 8 - 9 | 987 | 83 | |
| AK146362.1 | - | 8 - 9 | 1012 | 83 | ||
| AK150713.1 | - | 8 - 9 | 982 | 83 | ||
| AK151233.1 | - | 8 - 9 | 982 | 83 | ||
| AK151706.1 | - | 8 - 9 | 1005 | 83 | ||
| AK152691.1 | - | 8 - 9 | 83 | |||
| AK159822.1 | - | 8 - 9 | 979 | 83 | ||
| AK160569.1 | - | 8 - 9 | 979 | 83 | ||
| AK166667.1 | - | 8 - 9 | 981 | 83 | ||
| AK169129.1 | - | 8 - 9 | 1012 | 83 | ||
| AK169788.1 | - | 8 - 9 | 1012 | 83 | ||
| BC010314.1 | - | 8 - 9 | 946 | 2646679 | 83 | |
| BC052671.1 | - | 8 - 9 | 967 | 30058399 | 83 | |
| M33934.1 | - | 8 - 9 | 979 | 83 | ||
| M98333.1 | - | 8 - 9 | 913 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Impdh2
Genomic Map