Assay Details
| Gene Symbol: Apoh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013475.4 | NP_038503.4 | 5 - 6 | 646 | 58 | |
| GenBank mRNA | AA273732.1 | - | 2 - 3 | 261 | 58 | |
| AK010926.1 | - | 5 - 6 | 634 | 58 | ||
| BC011037.1 | - | 5 - 6 | 56 | |||
| BC019811.1 | - | 5 - 6 | 634 | 5125770 | 58 | |
| BC053338.1 | - | 5 - 6 | 622 | 5125963 | 58 | |
| BU756628.1 | - | 3 - 4 | 58 | |||
| D10056.1 | - | 5 - 6 | 620 | 58 | ||
| S70439.1 | - | 5 - 6 | 630 | 58 | ||
| Y11356.1 | - | 5 - 6 | 650 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Apoh
Genomic Map