Assay Details
| Gene Symbol: Emg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013536.2 | NP_038564.1 | 3 - 4 | 545 | 77 | |
| GenBank mRNA | AK081757.1 | - | 3 - 4 | 523 | 77 | |
| AK218607.1 | - | 1 - 2 | 66 | 77 | ||
| BC002004.1 | - | 3 - 4 | 465 | 3487480 | 77 | |
| CB948801.1 | - | 3 - 4 | 548 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Emg1
Genomic Map