Assay Details
| Gene Symbol: Emg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013536.2 | NP_038564.1 | 4 - 5 | 601 | 113 | |
| GenBank mRNA | AK081757.1 | - | 4 - 5 | 579 | 113 | |
| AK218607.1 | - | 2 - 3 | 122 | 113 | ||
| BC002004.1 | - | 4 - 5 | 521 | 3487480 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Emg1
Genomic Map