Assay Details
| Gene Symbol: Gstp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013541.1 | NP_038569.1 | 1 - 2 | 53 | 68 | |
| GenBank mRNA | BC002048.1 | - | 1 - 2 | 58 | 3593257 | 68 |
| BC061109.1 | - | 1 - 2 | 62 | 30322194 | 68 | |
| D30687.1 | - | 1 - 2 | 53 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC002048.1,BC061109.1,D30687.1,NM_013541.1 transcripts.
This assay is in the 5'UTR region of the BC002048.1,BC061109.1,D30687.1,NM_013541.1 transcripts.
Set Membership: |
|

Gstp1
Genomic Map